Please login to your online account to display your discounted pricing

(R)-(-)-4-Phenyl-2-oxazolidinone, 99%, ACROS Organics™


Quantity 5g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  303840050

 View more versions of this product

Catalog No. AC303840050

  • / Each
 

Description & Specifications

Chemical Identifiers

Assay ≥98.5%
Formula Weight 163.18
Physical Form Crystalline Powder
Quantity 5g
Color White
TSCA TSCA
Melting Point 129° to 132°C
Specific Rotation Condition − 49.50 (20.00°C c=2,CHCl3)
CAS Max % 100.0
Infrared Spectrum Authentic
CAS Min % 98.5
Packaging Glass bottle
Specific Rotation -48° to -51° (20°C, 589nm) (c=2, CHCl3)
Chemical Name or Material (R)-(-)-4-Phenyl-2-oxazolidinone
Assay Percent Range 99%
MDL Number MFCD00192393
Name Note 99%
CAS 90319-52-1
Molecular Formula C9H9NO2

Specifications

Chemical Name or Material (R)-(-)-4-Phenyl-2-oxazolidinone
Name Note 99%
CAS 90319-52-1
CAS Min % 98.5
CAS Max % 100.0
Assay ≥98.5%
Assay Percent Range 99%
Molecular Formula C9H9NO2
Formula Weight 163.18
MDL Number MFCD00192393