Please login to your online account to display your discounted pricing

(R)-(-)-4-Phenyl-2-oxazolidinone, 99%, ACROS Organics™


Quantity 1g
Packaging Glass bottle

Manufacturer: fisher scientific acros  303840010

 View more versions of this product

Catalog No. AC303840010

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥98.5%
Physical Form Crystalline Powder
Formula Weight 163.18
Quantity 1g
Color White
TSCA TSCA
Specific Rotation Condition − 49.50 (20.00°C c=2,CHCl3)
Melting Point 129° to 132°C
CAS Max % 100.0
Packaging Glass bottle
CAS Min % 98.5
Specific Rotation -48° to -51° (20°C, 589nm) (c=2, CHCl3)
Infrared Spectrum Authentic
Chemical Name or Material (R)-(-)-4-Phenyl-2-oxazolidinone
Molecular Formula C9H9NO2
MDL Number MFCD00192393
CAS 90319-52-1
Assay Percent Range 99%
Name Note 99%

Specifications

Chemical Name or Material (R)-(-)-4-Phenyl-2-oxazolidinone
Name Note 99%
CAS 90319-52-1
CAS Min % 98.5
CAS Max % 100.0
Assay ≥98.5%
Assay Percent Range 99%
Molecular Formula C9H9NO2
Formula Weight 163.18
MDL Number MFCD00192393