Please login to your online account to display your discounted pricing

(R)-(-)-4-Phenyl-2-oxazolidinone, 99%, ACROS Organics™

$310.16 - $310.16

Chemical Identifiers

Chemical Name or Material (R)-(-)-4-Phenyl-2-oxazolidinone
Name Note 99%
CAS 90319-52-1
CAS Min % 98.5
CAS Max % 100.0
Assay ≥98.5%
Assay Percent Range 99%
Molecular Formula C9H9NO2
Formula Weight 163.18
MDL Number MFCD00192393
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC303840010

  SDS
fisher scientific acros
303840010
Glass bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC303840050

  SDS
thermo fisher scientific
303840050
Glass bottle 5g Each for $310.16
Description & Specifications

Specifications

Assay ≥98.5%
Physical Form Crystalline Powder
Formula Weight 163.18
Quantity 1g
Color White
TSCA TSCA
Specific Rotation Condition − 49.50 (20.00°C c=2,CHCl3)
Melting Point 129° to 132°C
CAS Max % 100.0
Chemical Name or Material (R)-(-)-4-Phenyl-2-oxazolidinone
Specific Rotation -48° to -51° (20°C, 589nm) (c=2, CHCl3)
Infrared Spectrum Authentic
CAS Min % 98.5
Packaging Glass bottle
Name Note 99%
Assay Percent Range 99%
Molecular Formula C9H9NO2
MDL Number MFCD00192393
CAS 90319-52-1