Please login to your online account to display your discounted pricing

Alfa Aesar™ (R)-(-)-4-Phenyl-2-butanol, 99+%

Chemical Identifiers

Chemical Name or Material (R)-(-)-4-Phenyl-2-butanol
CAS 39516-03-5
Molecular Formula C10H14O
Formula Weight 150.22g/mol
MDL Number MFCD03093081
Beilstein 5248382
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL18885MD

 
alfa aesar™
L18885-MD
250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Boiling Point 239°C
Quantity 250mg
Flash Point >110°C (230°F)
Optical Rotation −14° (Neat)
Chemical Name or Material (R)-(-)-4-Phenyl-2-butanol
CAS 39516-03-5
Assay ≥99%
Molecular Formula C10H14O
Formula Weight 150.22g/mol
MDL Number MFCD03093081
Beilstein 5248382