missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

(R)-(+)-4-(Methoxymethyl)-1,3-dioxolan-2-one, TCI America™

$175.70 - $524.03

Chemical Identifiers

CAS 185836-34-4
Molecular Formula C5H8O4
Molecular Weight (g/mol) 132.115
MDL Number MFCD01321303
InChI Key DNSGQMOSYDHNHO-SCSAIBSYSA-N
Synonym (R)-(+)-4-(Methoxymethyl)-2-oxo-1,3-dioxolane, (R)-(+)-3-Methoxypropylene Carbonate
PubChem CID 13706498
IUPAC Name (4R)-4-(methoxymethyl)-1,3-dioxolan-2-one
SMILES COCC1COC(=O)O1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
M14515G View Documents TCI America
M14515G
5g Each for $175.70
 
M145125G View Documents TCI America
M145125G
25g Each for $524.03
 
Specifications

Chemical Identifiers

185836-34-4
132.115
DNSGQMOSYDHNHO-SCSAIBSYSA-N
13706498
COCC1COC(=O)O1
C5H8O4
MFCD01321303
(R)-(+)-4-(Methoxymethyl)-2-oxo-1,3-dioxolane, (R)-(+)-3-Methoxypropylene Carbonate
(4R)-4-(methoxymethyl)-1,3-dioxolan-2-one

Specifications

106°C
5g
C5H8O4
(R)-(+)-4-(Methoxymethyl)-2-oxo-1,3-dioxolane, (R)-(+)-3-Methoxypropylene Carbonate
COCC1COC(=O)O1
132.115
132.12
≥98.0% (GC)
Colorless
185836-34-4
MFCD01321303
DNSGQMOSYDHNHO-SCSAIBSYSA-N
(4R)-4-(methoxymethyl)-1,3-dioxolan-2-one
13706498
Clear Liquid at 20°C
(R)-(+)-4-(Methoxymethyl)-1,3-dioxolan-2-one
Safety and Handling

Safety and Handling

TSCA : No