missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ (R)-(+)-4-Hydroxy-2-pyrrolidinone, ≥97%

Manufacturer:  Alfa Aesar™ H66006MD

 View more versions of this product

Catalog No. AAH66006MD



Description

Description

Applications
(R)-(+)-4-Hydroxy-2-pyrrolidinone intermediate in the preparation of Oxiracetam. Also used as enantiomeric intermediate for the asymmetric single isomeric drugs including antitumor, anesthetic, antipasmodic, anti-inflammatory or anti-HIV activity products.

Solubility
Slightly soluble in water.

Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
Specifications

Specifications

(R)-(+)-4-Hydroxy-2-pyrrolidinone
155°C to 160°C
22677-21-0
MFCD00273364
2-pyrrolidinone, 4-hydroxy-, 4r, 4r-4-hydroxy-2-pyrrolidinone, 4r-4-hydroxypyrrolidin-2-one, r-+-4-hydroxy-2-pyrrolidinone, r-+-4-hydroxy-2-pyrrolidone, r-4-hydroxy-2-pyrrolidinone, r-4-hydroxy-2-pyrrolidone, r-4-hydroxy-pyrrolidin-2-one, r-4-hydroxypyrrolidin-2-one, rhpd
IOGISYQVOGVIEU-GSVOUGTGSA-N
(4R)-4-hydroxypyrrolidin-2-one
185505
Powder
White to Yellow
250mg
C4H7NO2
1524193
Slightly soluble in water.
C1C(CNC1=O)O
101.105
101.1
≥97%

Chemical Identifiers

22677-21-0
101.105
IOGISYQVOGVIEU-GSVOUGTGSA-N
185505
C1C(CNC1=O)O
C4H7NO2
MFCD00273364
2-pyrrolidinone, 4-hydroxy-, 4r, 4r-4-hydroxy-2-pyrrolidinone, 4r-4-hydroxypyrrolidin-2-one, r-+-4-hydroxy-2-pyrrolidinone, r-+-4-hydroxy-2-pyrrolidone, r-4-hydroxy-2-pyrrolidinone, r-4-hydroxy-2-pyrrolidone, r-4-hydroxy-pyrrolidin-2-one, r-4-hydroxypyrrolidin-2-one, rhpd
(4R)-4-hydroxypyrrolidin-2-one
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

Documents