Please login to your online account to display your discounted pricing

Alfa Aesar™ (R)-(-)-4-Fluorostyrene oxide, ChiPros 98+%, ee 99%

$327.80 - $327.80

Chemical Identifiers

Chemical Name or Material (R)-(-)-4-Fluorostyrene oxide
CAS 134356-73-3
Molecular Formula C8H7FO
Formula Weight 138.14g/mol
MDL Number MFCD03788740
UN Number UN1992
Beilstein 4350328
Synonym (R)-2-(4-Fluorophenyl)oxirane
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1958306

 
alfa aesar™
L19583-06
5g Each for $327.80
Description & Specifications

Specifications

Boiling Point 92°C (14mmHg)
Quantity 5g
Flash Point 77°C (171°F)
Optical Rotation −17° (c=1 in Chloroform)
Chemical Name or Material (R)-(-)-4-Fluorostyrene oxide
CAS 134356-73-3
Assay 99%
Assay Percent Notes ee 99%
Molecular Formula C8H7FO
Formula Weight 138.14g/mol
MDL Number MFCD03788740
UN Number UN1992
Beilstein 4350328
Synonym (R)-2-(4-Fluorophenyl)oxirane
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H331-H226-H351-H302-H312-H315-H319-H335
Toxic if inhaled.
Flammable liquid and vapor.
Suspected of causing cancer.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, TOXIC, N.O.S.
Safety and Handling

Danger

GHS P Statement
P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H331-H226-H351-H302-H312-H315-H319-H335
Toxic if inhaled.
Flammable liquid and vapor.
Suspected of causing cancer.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.