Please login to your online account to display your discounted pricing

(R)-4-Fluorophenylglycine, 95%, 98% e.e., ACROS Organics™


Quantity 5g
Packaging Glass bottle

Manufacturer: fisher scientific acros  363010050

 View more versions of this product

Catalog No. AC363010050

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Enantiomeric Excess 97.5% min. (HPLC)
Infrared Spectrum Authentic
Packaging Glass bottle
Quantity 5g
Color White to almost white
Chemical Name or Material (R)-4-Fluorophenylglycine
Name Note 95%, (98% e.e.)
CAS 93939-74-3
CAS Min % 94.0
CAS Max % 100.0
Physical Form White to almost white powder
Assay 94% min.
Assay Percent Range 95%
Molecular Formula C8 H8 F N O2
Formula Weight 169.15g/mol
MDL Number MFCD00042727
Health Hazard 1 HYGROSCOPIC
EINECS Number 300-363-8
TSCA TSCA

Specifications

Chemical Name or Material (R)-4-Fluorophenylglycine
Name Note 95%, (98% e.e.)
CAS 93939-74-3
CAS Min % 94.0
CAS Max % 100.0
Assay 94% min.
Assay Percent Range 95%
Molecular Formula C8 H8 F N O2
Formula Weight 169.15g/mol
MDL Number MFCD00042727
Accessories

HYGROSCOPIC

SDS & Safety Documents