Please login to your online account to display your discounted pricing

(R)-4-Fluorophenylglycine, 95%, 98% e.e., ACROS Organics™

Chemical Identifiers

Chemical Name or Material (R)-4-Fluorophenylglycine
Name Note 95%, (98% e.e.)
CAS 93939-74-3
CAS Min % 94.0
CAS Max % 100.0
Assay 94% min.
Assay Percent Range 95%
Molecular Formula C8 H8 F N O2
Formula Weight 169.15g/mol
MDL Number MFCD00042727
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC363010050

  SDS
fisher scientific acros
363010050
Glass bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Enantiomeric Excess 97.5% min. (HPLC)
Infrared Spectrum Authentic
Packaging Glass bottle
Quantity 5g
Color White to almost white
Chemical Name or Material (R)-4-Fluorophenylglycine
Name Note 95%, (98% e.e.)
CAS 93939-74-3
CAS Min % 94.0
CAS Max % 100.0
Physical Form White to almost white powder
Assay 94% min.
Assay Percent Range 95%
Molecular Formula C8 H8 F N O2
Formula Weight 169.15g/mol
MDL Number MFCD00042727
Health Hazard 1 HYGROSCOPIC
EINECS Number 300-363-8
TSCA TSCA
Safety and Handling

HYGROSCOPIC