Please login to your online account to display your discounted pricing

(R)-(+)-4-fluoro-α-methylbenzyl alcohol, ACROS Organics™


Quantity 1g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC375450010

 View more versions of this product

Catalog No. AC375450010

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥94%
Formula Weight 140.16
Physical Form Liquid
Quantity 1g
Color Clear colorless to pale yellow
Health Hazard 2 GHS H Statement:
Harmful if swallowed.
May cause respiratory irritation.
Causes serious eye irritation.
Harmful if inhaled.
Harmful in contact with skin.
Causes skin irritation.
MDL Number MFCD03092997
Chemical Name or Material (R)-(+)-4-Fluoro-α-methylbenzyl alcohol
Packaging Glass bottle
Synonym (R)-(+)-1-(4-Fluorophenyl)ethanol
Enantiomeric Excess 97.5% min. (GC)
Infrared Spectrum Authentic
Linear Formula CH3CH(OH)C6H4F
Health Hazard 1 GHS Signal Word: Warning
CAS 101219-68-5
Molecular Formula C8H9FO
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Specifications

Chemical Name or Material (R)-(+)-4-Fluoro-α-methylbenzyl alcohol
Assay ≥94%
Molecular Formula C8H9FO
Linear Formula CH3CH(OH)C6H4F
Formula Weight 140.16
MDL Number MFCD03092997
Synonym (R)-(+)-1-(4-Fluorophenyl)ethanol
Accessories

GHS Signal Word: Warning

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Harmful if swallowed.
May cause respiratory irritation.
Causes serious eye irritation.
Harmful if inhaled.
Harmful in contact with skin.
Causes skin irritation.

SDS & Safety Documents