Please login to your online account to display your discounted pricing

(R)-(+)-4-fluoro-α-methylbenzyl alcohol, ACROS Organics™

$51.94 - $51.94

Chemical Identifiers

Chemical Name or Material (R)-(+)-4-Fluoro-α-methylbenzyl alcohol
Assay ≥94%
Molecular Formula C8H9FO
Linear Formula CH3CH(OH)C6H4F
Formula Weight 140.16
MDL Number MFCD03092997
Synonym (R)-(+)-1-(4-Fluorophenyl)ethanol
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC375450010

  SDS
thermo fisher scientific
AC375450010
Glass bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC375452500

  SDS
thermo fisher scientific
AC375452500
Glass Ampule with Screw-cap 250mg Each for $51.94
Description & Specifications

Specifications

Assay ≥94%
Physical Form Liquid
Formula Weight 140.16
Quantity 1g
Color Clear colorless to pale yellow
Chemical Name or Material (R)-(+)-4-Fluoro-α-methylbenzyl alcohol
Enantiomeric Excess 97.5% min. (GC)
Linear Formula CH3CH(OH)C6H4F
MDL Number MFCD03092997
CAS 101219-68-5
Synonym (R)-(+)-1-(4-Fluorophenyl)ethanol
Health Hazard 1 GHS Signal Word: Warning
Health Hazard 2 GHS H Statement:
Harmful if swallowed.
May cause respiratory irritation.
Causes serious eye irritation.
Harmful if inhaled.
Harmful in contact with skin.
Causes skin irritation.
Infrared Spectrum Authentic
Packaging Glass bottle
Molecular Formula C8H9FO
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Safety and Handling

GHS Signal Word: Warning

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Harmful if swallowed.
May cause respiratory irritation.
Causes serious eye irritation.
Harmful if inhaled.
Harmful in contact with skin.
Causes skin irritation.