Please login to your online account to display your discounted pricing

Alfa Aesar™ (R)-(+)-4-(Diphenylmethyl)-2-oxazolidinone, 98%

$321.72 - $321.72

Chemical Identifiers

Chemical Name or Material (R)-(+)-4-(Diphenylmethyl)-2-oxazolidinone
CAS 173604-33-6
Molecular Formula C16H15NO2
Formula Weight 253.3
MDL Number MFCD01863565
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH5107403

  SDS
alfa aesar™
H5107403
1g Each for $321.72
Description & Specifications

Specifications

Formula Weight 253.3
Chemical Name or Material (R)-(+)-4-(Diphenylmethyl)-2-oxazolidinone
Assay 98%
Molecular Formula C16H15NO2
MDL Number MFCD01863565
CAS 173604-33-6
Quantity 1g

Applications
(R)-(+)-4-(Diphenylmethyl)-2-oxazolidinone is used as organic chemical synthesis intermediate.

Solubility
Slightly soluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.