Please login to your online account to display your discounted pricing

(R)-(+)-4-(Chloromethyl)-2,2-dimethyl-1,3-dioxolane, 98%, ACROS Organics™


Quantity 1g

Manufacturer: thermo fisher scientific  AC399220010

 View more versions of this product

Catalog No. AC399220010

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Quantity 1g
Chemical Name or Material (R)-(+)-4-(Chloromethyl)-2, 2-dimethyl-1, 3-dioxolane
Assay Percent Range 98%
Molecular Formula C6H11ClO2
MDL Number MFCD00273371
CAS 57044-24-3

Specifications

Chemical Name or Material (R)-(+)-4-(Chloromethyl)-2, 2-dimethyl-1, 3-dioxolane
CAS 57044-24-3
Assay Percent Range 98%
Molecular Formula C6H11ClO2
MDL Number MFCD00273371