missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

(R)-(+)-4-(Chloromethyl)-2,2-dimethyl-1,3-dioxolane, 98%, ACROS Organics™

Chemical Identifiers

CAS 57044-24-3
Assay Percent Range 98%
Molecular Formula C6H11ClO2
MDL Number MFCD00273371
Chemical Name or Material (R)-(+)-4-(Chloromethyl)-2, 2-dimethyl-1, 3-dioxolane
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AC399220010 SDS Thermo Fisher Scientific
AC399220010
1g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC399220050 SDS Thermo Fisher Scientific
AC399220050
5g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

57044-24-3
C6H11ClO2
(R)-(+)-4-(Chloromethyl)-2, 2-dimethyl-1, 3-dioxolane
98%
MFCD00273371

Specifications

57044-24-3
1g
MFCD00273371
98%
C6H11ClO2
(R)-(+)-4-(Chloromethyl)-2, 2-dimethyl-1, 3-dioxolane