missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

(R)-(-)-4-Benzyl-3-propionyl-2-oxazolidinone, TCI America™

$102.36 - $990.02

Chemical Identifiers

CAS 131685-53-5
Molecular Formula C13H15NO3
Molecular Weight (g/mol) 233.267
MDL Number MFCD00269687
InChI Key WHOBYFHKONUTMW-LLVKDONJSA-N
PubChem CID 10966403
IUPAC Name (4R)-4-benzyl-3-propanoyl-1,3-oxazolidin-2-one
SMILES CCC(=O)N1C(COC1=O)CC2=CC=CC=C2
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
B21651G View Documents TCI America
B21651G
1g Each for $102.36
 
B216525G View Documents TCI America
B216525G
25g Each for $990.02
 
B21655G View Documents TCI America
B21655G
5g Each for $293.34
 
Specifications

Chemical Identifiers

131685-53-5
233.267
WHOBYFHKONUTMW-LLVKDONJSA-N
(4R)-4-benzyl-3-propanoyl-1,3-oxazolidin-2-one
C13H15NO3
MFCD00269687
10966403
CCC(=O)N1C(COC1=O)CC2=CC=CC=C2

Specifications

White-Yellow
1g
C13H15NO3
WHOBYFHKONUTMW-LLVKDONJSA-N
(4R)-4-benzyl-3-propanoyl-1,3-oxazolidin-2-one
10966403
Crystal-Powder at 20°C
(R)-(-)-4-Benzyl-3-propionyl-2-oxazolidinone
45°C
131685-53-5
MFCD00269687
CCC(=O)N1C(COC1=O)CC2=CC=CC=C2
233.267
233.27
≥98.0% (GC)
Safety and Handling

Safety and Handling

TSCA : No