Please login to your online account to display your discounted pricing

Alfa Aesar™ (R)-(-)-4-(3-Indolylmethyl)-2-oxazolidinone, 98%

$270.92 - $270.92

Chemical Identifiers

Chemical Name or Material (R)-(-)-4-(3-Indolylmethyl)-2-oxazolidinone
CAS 157636-81-2
Molecular Formula C12H12N2O2
Formula Weight 216.24
MDL Number MFCD01863583
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH5107503

 
alfa aesar™
H5107503
1g Each for $270.92
Description & Specifications

Specifications

Formula Weight 216.24
Chemical Name or Material (R)-(-)-4-(3-Indolylmethyl)-2-oxazolidinone
Assay 98%
Molecular Formula C12H12N2O2
MDL Number MFCD01863583
CAS 157636-81-2
Quantity 1g
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.