Please login to your online account to display your discounted pricing

Alfa Aesar™ (R)-4,12-Dibromo[2.2]paracyclophane, 98%, ee 98%

Chemical Identifiers

Chemical Name or Material (R)-4,12-Dibromo[2.2]paracyclophane
Molecular Formula C16H14Br2
Formula Weight 366.10g/mol
MDL Number MFCD09953457
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2580903

 
alfa aesar™
H25809-03
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Melting Point 202° to 204°C
Quantity 1g
Optical Rotation −128° (c=1 in Chloroform)
Chemical Name or Material (R)-4,12-Dibromo[2.2]paracyclophane
Assay 98%
Assay Percent Notes ee 98%
Molecular Formula C16H14Br2
Formula Weight 366.10g/mol
MDL Number MFCD09953457