Please login to your online account to display your discounted pricing

Alfa Aesar™ (R)-(-)-4,12-Bis(di-3,5-xylylphosphino)[2.2]paracyclophane, CTH-(R)-3,5-xylyl-PHANEPHOS

Asymmetric hydrogenation

$489.18 - $489.18

Chemical Identifiers

Chemical Name or Material (R)- (-)-4,12-Bis (di-3,5-xylylphosphino)[2.2]paracyclophane
CAS 325168-89-6
Name Note CTH-(R)-3,5-xylyl-PHANEPHOS
Molecular Formula C48H50P2
Formula Weight 688.87g/mol
MDL Number MFCD03840578
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA4461502

 
alfa aesar™
44615-02
0.5g Each for $489.18
Description & Specifications

Specifications

Quantity 0.5g
Color White
Quantity 0.5g
Odor Odorless
Optical Rotation −98° (c=1 in dichloromethane)
Chemical Name or Material (R)- (-)-4,12-Bis (di-3,5-xylylphosphino)[2.2]paracyclophane
Name Note CTH-(R)-3,5-xylyl-PHANEPHOS
CAS 325168-89-6
Physical Form Powder
Molecular Formula C48H50P2
Formula Weight 688.87g/mol
MDL Number MFCD03840578
Storage Note 1
  • Packaged under Argon

  • Packaged under Argon

Safety and Handling

  • Packaged under Argon