Please login to your online account to display your discounted pricing

(R)-4-(1-Hydroxyethyl)pyridine 99+%, (99% ee), ACROS Organics™

Chemical Identifiers

Chemical Name or Material (R)-4-(1-Hydroxyethyl)pyridine
CAS 27854-88-2
CAS Min % 99.0
CAS Max % 100.0
Assay ≥99.0%
Assay Percent Range 99+%
Molecular Formula C7H9NO
Formula Weight 123.15
MDL Number MFCD00077865
Synonym (R)-(+)-1-(4-Pyridyl)ethanol
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC393360010

  SDS
thermo fisher scientific
AC393360010
Glass bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC393362500

  SDS
thermo fisher scientific
AC393362500
Glass bottle 250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥99.0%
Physical Form Crystalline Powder or Needles
Formula Weight 123.15
Quantity 1g
Color White
Specific Rotation Condition +58.50° (20°C c=1,chloroform)
Melting Point 67° to 70°C
CAS Min % 99.0
CAS Max % 100.0
Synonym (R)-(+)-1-(4-Pyridyl)ethanol
Chemical Name or Material (R)-4-(1-Hydroxyethyl)pyridine
Assay Percent Range 99+%
Molecular Formula C7H9NO
MDL Number MFCD00077865
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Enantiomeric Excess 99.0% min.
Infrared Spectrum Authentic
Packaging Glass bottle
Specific Rotation +53° to +59° (20°C, 589nm) (c=1, CHCl3)
CAS 27854-88-2
Health Hazard 2 GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
Harmful if swallowed.
May cause respiratory irritation.
Safety and Handling

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
Harmful if swallowed.
May cause respiratory irritation.