Please login to your online account to display your discounted pricing

Alfa Aesar™ (R)-4-(1-Aminoethyl)phenol, 97%

Chemical Identifiers

Chemical Name or Material (R)-4-(1-Aminoethyl)phenol
CAS 134855-89-3
Formula Weight 137.18
MDL Number MFCD03844647
UN Number UN3259
Synonym (R)-1-(4-Hydroxyphenyl)ethylamine
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6300103

 
alfa aesar™
H6300103
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6300106

 
alfa aesar™
H6300106
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH63001MD

 
alfa aesar™
H63001MD
250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 137.18
Chemical Name or Material (R)-4-(1-Aminoethyl)phenol
Physical Form Solid
Assay 97%
MDL Number MFCD03844647
CAS 134855-89-3
UN Number UN3259
Synonym (R)-1-(4-Hydroxyphenyl)ethylamine
Health Hazard 1 Danger
Color White
Quantity 1g
Health Hazard 2 GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Health Hazard 3 GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Storage Note 1 2° to 8°C
DOT Information Transport Hazard Class: 8; Packaging Group: III; Proper Shipping Name: AMINES, SOLID, CORROSIVE, N.O.S.
Safety and Handling

Danger

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

2° to 8°C