missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
(R)-3-Pyrrolidinol hydrochloride, 98%, ACROS Organics™
Manufacturer: Thermo Fisher Scientific AC442100010
Specifications
(R)-3-Pyrrolidinol hydrochloride | |
-7.6 | |
1g | |
97.5% min. (Argentometry) | |
3r-3-hydroxypyrrolidine hydrochloride, 3r-pyrrolidin-3-ol hydrochloride, 3r-pyrrolidinol hydrochloride, r-+-3-pyrrolidinol hydrochloride, r---3-pyrrolidinol hydrochloride, r-3-hydroxypyrrolidine hcl, r-3-hydroxypyrrolidine hydrochloride, r-3-pyrrolidinol hydrochloride, r-pyrrolidin-3-ol hydrochloride | |
C1CNCC1O.Cl | |
123.58 | |
123.58 |
Glass Bottle | |
102°C to 107.5°C | |
104706-47-0 | |
C4H9NO·ClH | |
QPMSJEFZULFYTB-PGMHMLKASA-N | |
(3R)-pyrrolidin-3-ol;hydrochloride | |
2759336 | |
98% |
Chemical Identifiers
104706-47-0 | |
123.58 | |
3r-3-hydroxypyrrolidine hydrochloride, 3r-pyrrolidin-3-ol hydrochloride, 3r-pyrrolidinol hydrochloride, r-+-3-pyrrolidinol hydrochloride, r---3-pyrrolidinol hydrochloride, r-3-hydroxypyrrolidine hcl, r-3-hydroxypyrrolidine hydrochloride, r-3-pyrrolidinol hydrochloride, r-pyrrolidin-3-ol hydrochloride | |
(3R)-pyrrolidin-3-ol;hydrochloride |
C4H9NO·ClH | |
QPMSJEFZULFYTB-PGMHMLKASA-N | |
2759336 | |
C1CNCC1O.Cl |
Safety and Handling
GHS Signal Word: Warning