missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
(R)-3-Pyrrolidinol hydrochloride, 98%, Thermo Scientific™
Chemical Identifiers
CAS | 104706-47-0 |
---|---|
Molecular Formula | C4H9NO·ClH |
Molecular Weight (g/mol) | 123.58 |
InChI Key | QPMSJEFZULFYTB-PGMHMLKASA-N |
Synonym | r-3-hydroxypyrrolidine hydrochloride, r---3-pyrrolidinol hydrochloride, 3r-pyrrolidin-3-ol hydrochloride, r-pyrrolidin-3-ol hydrochloride, r-3-hydroxypyrrolidine hcl, r-3-pyrrolidinol hydrochloride, 3r-3-hydroxypyrrolidine hydrochloride, 3r-pyrrolidinol hydrochloride, r-+-3-pyrrolidinol hydrochloride |
PubChem CID | 2759336 |
IUPAC Name | (3R)-pyrrolidin-3-ol;hydrochloride |
SMILES | C1CNCC1O.Cl |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AC442100010
|
Thermo Scientific Chemicals
442100010 |
1g |
N/A
|
N/A | |||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Chemical Identifiers
104706-47-0 | |
123.58 | |
r-3-hydroxypyrrolidine hydrochloride, r---3-pyrrolidinol hydrochloride, 3r-pyrrolidin-3-ol hydrochloride, r-pyrrolidin-3-ol hydrochloride, r-3-hydroxypyrrolidine hcl, r-3-pyrrolidinol hydrochloride, 3r-3-hydroxypyrrolidine hydrochloride, 3r-pyrrolidinol hydrochloride, r-+-3-pyrrolidinol hydrochloride | |
(3R)-pyrrolidin-3-ol;hydrochloride |
C4H9NO·ClH | |
QPMSJEFZULFYTB-PGMHMLKASA-N | |
2759336 | |
C1CNCC1O.Cl |
Specifications
104706-47-0 | |
Glass Bottle | |
1g | |
r-3-hydroxypyrrolidine hydrochloride, r---3-pyrrolidinol hydrochloride, 3r-pyrrolidin-3-ol hydrochloride, r-pyrrolidin-3-ol hydrochloride, r-3-hydroxypyrrolidine hcl, r-3-pyrrolidinol hydrochloride, 3r-3-hydroxypyrrolidine hydrochloride, 3r-pyrrolidinol hydrochloride, r-+-3-pyrrolidinol hydrochloride | |
C1CNCC1O.Cl | |
123.58 | |
123.58 | |
(R)-3-Pyrrolidinol hydrochloride |
97.5% min. (Argentometry) | |
C4H9NO·ClH | |
-7.6 | |
QPMSJEFZULFYTB-PGMHMLKASA-N | |
(3R)-pyrrolidin-3-ol;hydrochloride | |
2759336 | |
98% |
Safety and Handling
GHS Signal Word: Warning
RUO â Research Use Only