Please login to your online account to display your discounted pricing

(R)-(+)-3-Pyrrolidinol, 98%, ACROS Organics™


Quantity 5g
Packaging Glass bottle

Manufacturer: fisher scientific acros  344570050

 View more versions of this product

Catalog No. AC344570050

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥97.5%
Physical Form Viscous Liquid
Formula Weight 87.12
Quantity 5g
Color Clear yellow to orange-brown
Specific Rotation Condition +6° (20°C C=3.5,CH3OH)
Density 1.048g/mL
Chemical Name or Material (R)-(+)-3-Pyrrolidinol
Infrared Spectrum Authentic
Packaging Glass bottle
Refractive Index 1.4870 to 1.4910 (20°C, 589nm)
Specific Rotation +4.5° to +7.5° (20°C, 589nm) (c=3.5, CH3OH)
Storage Information Product darkens on storage
Synonym (R)-3-Hydroxypyrrolidine
Health Hazard 2 GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Assay Percent Range 98%
Molecular Formula C4H9NO
MDL Number MFCD00145220
CAS 2799-21-5

Specifications

Chemical Name or Material (R)-(+)-3-Pyrrolidinol
CAS 2799-21-5
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C4H9NO
Formula Weight 87.12
MDL Number MFCD00145220
Synonym (R)-3-Hydroxypyrrolidine
Accessories

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.

SDS & Safety Documents