missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

(R)-(-)-3-Piperidinecarboxylic Acid, TCI America™

$51.95 - $154.31

Chemical Identifiers

CAS 25137-00-2
Molecular Formula C6H11NO2
Molecular Weight (g/mol) 129.159
MDL Number MFCD01630787
InChI Key XJLSEXAGTJCILF-RXMQYKEDSA-N
Synonym r-piperidine-3-carboxylic acid, r---nipecotic acid, r-nipecotic acid, 3r-piperidine-3-carboxylic acid, r---3-piperidinecarboxylic acid, --nipecotic acid, d--nipecotic acid, 3r---piperidine-3-carboxylic acid, r-3-piperidinecarboxylic acid, 3r-nipecotic acid
PubChem CID 1796510
ChEBI CHEBI:221278
IUPAC Name (3R)-piperidine-3-carboxylic acid
SMILES C1CC(CNC1)C(=O)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
N06545G View Documents TCI America
N06545G
5g Each for $154.31
 
N06541G View Documents TCI America
N06541G
1g Each for $51.95
 
Specifications

Chemical Identifiers

25137-00-2
129.159
XJLSEXAGTJCILF-RXMQYKEDSA-N
1796510
(3R)-piperidine-3-carboxylic acid
C6H11NO2
MFCD01630787
r-piperidine-3-carboxylic acid, r---nipecotic acid, r-nipecotic acid, 3r-piperidine-3-carboxylic acid, r---3-piperidinecarboxylic acid, --nipecotic acid, d--nipecotic acid, 3r---piperidine-3-carboxylic acid, r-3-piperidinecarboxylic acid, 3r-nipecotic acid
CHEBI:221278
C1CC(CNC1)C(=O)O

Specifications

White
25137-00-2
MFCD01630787
XJLSEXAGTJCILF-RXMQYKEDSA-N
(3R)-piperidine-3-carboxylic acid
1796510
129.16
≥98.0% (T)
5g
C6H11NO2
r-piperidine-3-carboxylic acid, r---nipecotic acid, r-nipecotic acid, 3r-piperidine-3-carboxylic acid, r---3-piperidinecarboxylic acid, --nipecotic acid, d--nipecotic acid, 3r---piperidine-3-carboxylic acid, r-3-piperidinecarboxylic acid, 3r-nipecotic acid
C1CC(CNC1)C(=O)O
129.159
CHEBI:221278
Crystal-Powder at 20°C
(R)-(-)-3-Piperidinecarboxylic Acid
Safety and Handling

Safety and Handling

TSCA : No