missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
(R)-(-)-3-Piperidinamine dihydrochloride, 97%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 436412500
Specifications
(R)-(-)-3-Piperidinamine dihydrochloride | |
96.0 | |
33.5 to 35.9% C, 15.5 to 16.8% N | |
97% | |
250mg | |
r-3-aminopiperidine dihydrochloride, r-piperidin-3-amine dihydrochloride, r---3-aminopiperidine dihydrochloride, r-3-piperidinamine dihydrochloride, 3r-piperidin-3-amine dihydrochloride, r-3-amino-piperidine dihydrochloride, r---3-aminopiperidine 2hcl, r-3-aminopiperidine 2hcl, r-3-amino piperidine dihcl | |
C1CC(CNC1)N.Cl.Cl | |
173.09 | |
173.09 |
334618-23-4 | |
100.0 | |
Authentic | |
C5H12N2·2ClH | |
-1° to -3° (20°C, 589nm) (c=1, water) | |
GGPNYXIOFZLNKW-ZJIMSODOSA-N | |
(3R)-piperidin-3-amine;dihydrochloride | |
16211333 | |
97% |
Chemical Identifiers
334618-23-4 | |
173.09 | |
r-3-aminopiperidine dihydrochloride, r-piperidin-3-amine dihydrochloride, r---3-aminopiperidine dihydrochloride, r-3-piperidinamine dihydrochloride, 3r-piperidin-3-amine dihydrochloride, r-3-amino-piperidine dihydrochloride, r---3-aminopiperidine 2hcl, r-3-aminopiperidine 2hcl, r-3-amino piperidine dihcl | |
(3R)-piperidin-3-amine;dihydrochloride |
C5H12N2·2ClH | |
GGPNYXIOFZLNKW-ZJIMSODOSA-N | |
16211333 | |
C1CC(CNC1)N.Cl.Cl |
Safety and Handling
GHS H Statement
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
Harmful if inhaled.
May cause respiratory irritation.
GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED.
IF INHALED: Remove victim
GHS Signal Word: Warning
RUO â Research Use Only