missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

(R)-3-Phenylcyclohexanone, TCI America™

Manufacturer:  TCI America P1499100MG

 View more versions of this product

Catalog No. P1499100MG



Specifications

Specifications

(R)-3-Phenylcyclohexanone
100mg
C12H14O
C1CC(CC(=O)C1)C2=CC=CC=C2
174.243
174.24
≥97.0% (GC)
Yellow
34993-51-6
CJAUDSQXFVZPTO-LLVKDONJSA-N
(3R)-3-phenylcyclohexan-1-one
11041212
Clear Liquid at 20°C

Chemical Identifiers

34993-51-6
174.243
11041212
C1CC(CC(=O)C1)C2=CC=CC=C2
C12H14O
CJAUDSQXFVZPTO-LLVKDONJSA-N
(3R)-3-phenylcyclohexan-1-one
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Refrigerator

Documents