Please login to your online account to display your discounted pricing

Alfa Aesar™ (R)-(-)-3-Methyl-2-butyl isocyanate, 97%

Chemical Identifiers

Chemical Name or Material (R)-(-)-3-Methyl-2-butyl isocyanate
CAS 745783-84-0
Molecular Formula C6H11NO
Formula Weight 113.16
MDL Number MFCD05664086
UN Number UN3080
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL2024103

  SDS
alfa aesar™
L2024103
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 113.16
Health Hazard 2 GHS H Statement
H330-H334-H226-H302-H315-H319-H335
Fatal if inhaled.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Flammable liquid and vapor.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Chemical Name or Material (R)-(-)-3-Methyl-2-butyl isocyanate
Molecular Formula C6H11NO
CAS 745783-84-0
Assay 97%
Health Hazard 1 Danger
MDL Number MFCD05664086
Health Hazard 3 GHS P Statement
P260-P280h-P305+P351+P338-P309-P310
Do not breathe dust/fume/gas/mist/vapors/spray.
Wear protective gloves/clothing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
Boiling Point 124°C
Recommended Storage 2° to 8°C
Flash Point 28°C (82°F)
Optical Rotation −29° (c=1 in Dichloromethane)
UN Number UN3080
DOT Information Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: ISOCYANATES, TOXIC, FLAMMABLE, N.O.S.
Quantity 1g
Safety and Handling

Danger

GHS P Statement
P260-P280h-P305+P351+P338-P309-P310
Do not breathe dust/fume/gas/mist/vapors/spray.
Wear protective gloves/clothing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.

GHS H Statement
H330-H334-H226-H302-H315-H319-H335
Fatal if inhaled.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Flammable liquid and vapor.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

2° to 8°C