missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ (R)-3-Isopropyl-2,5-dimethoxy-3,6-dihydropyrazine, 98%

Manufacturer:  Alfa Aesar™ H56077XF

 View more versions of this product

Catalog No. AAH56077XF



Description

Description

Applications
(R)-3-Isopropyl-2,5-dimethoxy-3,6-dihydropyrazineChiral auxiliary for the synthesis of α-amino acids. It is an inhibitor with antitumor activity.

Solubility
Soluble in Chloroform and Ethanol

Notes
Store at -20°C. Keep away from oxidizing agents. Hygroscopic. Keep under inert atmosphere.
Specifications

Specifications

(R)-3-Isopropyl-2,5-dimethoxy-3,6-dihydropyrazine
85°C (185°F)
1mL
C9H16N2O2
3542790
Soluble in Chloroform and Ethanol
CC(C)C1C(=NCC(=N1)OC)OC
184.239
184.24
1.028
−102° (c=1 in Ethanol)
109838-85-9
MFCD00040565
2r---2,5-dihydro-3,6-dimethoxy-2-isopropylpyrazine
FCFWEOGTZZPCTO-MRVPVSSYSA-N
(2R)-3,6-dimethoxy-2-propan-2-yl-2,5-dihydropyrazine
736066
98%

Chemical Identifiers

109838-85-9
184.239
FCFWEOGTZZPCTO-MRVPVSSYSA-N
736066
CC(C)C1C(=NCC(=N1)OC)OC
C9H16N2O2
MFCD00040565
2r---2,5-dihydro-3,6-dimethoxy-2-isopropylpyrazine
(2R)-3,6-dimethoxy-2-propan-2-yl-2,5-dihydropyrazine
Safety and Handling

Safety and Handling

GHS H Statement
H227
Combustible liquid.

P210-P280-P370+P378q-P501c

H227

TSCA : No

Recommended Storage : Ambient temperatures

Documents