missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

(R)-3-Hydroxytetrahydrofuran, TCI America™

$165.00 - $574.45

Chemical Identifiers

CAS 86087-24-3
Molecular Formula C4H8O2
Molecular Weight (g/mol) 88.106
MDL Number MFCD00067101
InChI Key XDPCNPCKDGQBAN-SCSAIBSYSA-N
Synonym r---3-hydroxytetrahydrofuran, r-tetrahydrofuran-3-ol, r-3-hydroxytetrahydrofuran, 3r-oxolan-3-ol, 3r-tetrahydrofuran-3-ol, r-+-3-hydroxytetrahydrofuran, r-tetrahydro-furan-3-ol, 3-furanol, tetrahydro-, 3r, pubchem7071, 2-furanyl,tetrahydro-3-hydroxy-, r-9ci
PubChem CID 641512
IUPAC Name (3R)-oxolan-3-ol
SMILES C1COCC1O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
H13325G View Documents TCI America
H13325G
5g Each for $574.45
 
H13321G View Documents TCI America
H13321G
1g Each for $165.00
 
Specifications

Chemical Identifiers

86087-24-3
88.106
XDPCNPCKDGQBAN-SCSAIBSYSA-N
641512
C1COCC1O
C4H8O2
MFCD00067101
r---3-hydroxytetrahydrofuran, r-tetrahydrofuran-3-ol, r-3-hydroxytetrahydrofuran, 3r-oxolan-3-ol, 3r-tetrahydrofuran-3-ol, r-+-3-hydroxytetrahydrofuran, r-tetrahydro-furan-3-ol, 3-furanol, tetrahydro-, 3r, pubchem7071, 2-furanyl,tetrahydro-3-hydroxy-, r-9ci
(3R)-oxolan-3-ol

Specifications

80°C
5g
C4H8O2
r---3-hydroxytetrahydrofuran, r-tetrahydrofuran-3-ol, r-3-hydroxytetrahydrofuran, 3r-oxolan-3-ol, 3r-tetrahydrofuran-3-ol, r-+-3-hydroxytetrahydrofuran, r-tetrahydro-furan-3-ol, 3-furanol, tetrahydro-, 3r, pubchem7071, 2-furanyl,tetrahydro-3-hydroxy-, r-9ci
C1COCC1O
88.106
88.11
≥98.0% (GC)
Colorless
86087-24-3
MFCD00067101
XDPCNPCKDGQBAN-SCSAIBSYSA-N
(3R)-oxolan-3-ol
641512
Clear Liquid at 20°C
(R)-3-Hydroxytetrahydrofuran
Safety and Handling

Safety and Handling

TSCA : No