missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ (R)-(-)-3-Hydroxytetrahydrofuran, 98%

Manufacturer:  Alfa Aesar™ H56307MD

 View more versions of this product

Catalog No. AAH56307MD



Specifications

Specifications

(R)-(-)-3-Hydroxytetrahydrofuran
−18° (c=2.4 in Methanol)
250mg
C4H8O2
r---3-hydroxytetrahydrofuran, r-tetrahydrofuran-3-ol, r-3-hydroxytetrahydrofuran, 3r-oxolan-3-ol, 3r-tetrahydrofuran-3-ol, r-+-3-hydroxytetrahydrofuran, r-tetrahydro-furan-3-ol, 3-furanol, tetrahydro-, 3r, pubchem7071, 2-furanyl,tetrahydro-3-hydroxy-, r-9ci
C1COCC1O
88.106
88.11
82°C (180°F)
181°C
86087-24-3
MFCD00067101
XDPCNPCKDGQBAN-SCSAIBSYSA-N
(3R)-oxolan-3-ol
641512
98%

Chemical Identifiers

86087-24-3
88.106
XDPCNPCKDGQBAN-SCSAIBSYSA-N
641512
C1COCC1O
C4H8O2
MFCD00067101
r---3-hydroxytetrahydrofuran, r-tetrahydrofuran-3-ol, r-3-hydroxytetrahydrofuran, 3r-oxolan-3-ol, 3r-tetrahydrofuran-3-ol, r-+-3-hydroxytetrahydrofuran, r-tetrahydro-furan-3-ol, 3-furanol, tetrahydro-, 3r, pubchem7071, 2-furanyl,tetrahydro-3-hydroxy-, r-9ci
(3R)-oxolan-3-ol
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.

GHS P Statement
P210-P261-P280-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

Documents