Please login to your online account to display your discounted pricing

(R)-(+)-3-Hydroxypiperidine hydrochloride, 98%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material (R)-(+)-3-Hydroxypiperidine hydrochloride
CAS 198976-43-1
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Molecular Formula C5H12ClNO
Formula Weight 137.61
MDL Number MFCD00192221
Synonym (R)-(+)-3-Piperidinol hydrochloride
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AC434470050

  SDS
thermo fisher scientific
AC434470050
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC434470010

  SDS
thermo fisher scientific
AC434470010
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Crystalline Powder
Formula Weight 137.61
Quantity 5g
Color White
Melting Point 133° to 139°C
CAS Max % 100.0
Additional Information May contain up to 3% water
Assay Percent Range 98%
Chemical Name or Material (R)-(+)-3-Hydroxypiperidine hydrochloride
Molecular Formula C5H12ClNO
MDL Number MFCD00192221
CAS 198976-43-1
CAS Min % 97.5
Elemental Analysis 42.18 to 44.73%, 9.92 to 10.43%
Specific Rotation +5° to +8° (20°C, 589nm) (c=1, methanol)
Synonym (R)-(+)-3-Piperidinol hydrochloride
Health Hazard 2 GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Infrared Spectrum Authentic
Safety and Handling

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.