Please login to your online account to display your discounted pricing

Alfa Aesar™ (R)-(+)-3-(Dimethylamino)pyrrolidine, 95%

Chemical Identifiers

Chemical Name or Material (R)-(+)-3-(Dimethylamino)pyrrolidine
CAS 132958-72-6
Molecular Formula C6H14N2
Formula Weight 114.19g/mol
MDL Number MFCD00191347
UN Number UN2733
Synonym (3R)-N,N-Dimethyl-3-pyrrolidinamine
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH26251MD

 
alfa aesar™
H26251-MD
250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Boiling Point 166°C
Quantity 250mg
Flash Point 51°C (124°F)
Chemical Name or Material (R)-(+)-3-(Dimethylamino)pyrrolidine
CAS 132958-72-6
Assay 95%
Molecular Formula C6H14N2
Formula Weight 114.19g/mol
MDL Number MFCD00191347
UN Number UN2733
Synonym (3R)-N,N-Dimethyl-3-pyrrolidinamine
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H314-H318-H226
Causes severe skin burns and eye damage.
Causes serious eye damage.
Flammable liquid and vapour.
Health Hazard 3 GHS P Statement
P210-P260-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Recommended Storage 2° to 8°C
DOT Information Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: AMINES, FLAMMABLE, CORROSIVE, N.O.S.
Safety and Handling

Danger

GHS P Statement
P210-P260-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H314-H318-H226
Causes severe skin burns and eye damage.
Causes serious eye damage.
Flammable liquid and vapour.

2° to 8°C