Please login to your online account to display your discounted pricing

(R)-3-Carboxy-4-hydroxyphenylglycine, Tocris


Ionotropic glutamate receptor antagonist

Quantity 10mg

Manufacturer: r & d systems  RDS032810

 View more versions of this product

Catalog No. 032810

  • / Each
 

Description & Specifications

Chemical Identifiers

Quantity 10mg
Formula Weight 211.17
Chemical Name or Material (R)-3-Carboxy-4-hydroxyphenylglycine
Assay Percent Range >98%
CAS 13861-03-5
Molecular Formula C9H9NO5
Recommended Storage Store at Room Temperature

Specifications

Chemical Name or Material (R)-3-Carboxy-4-hydroxyphenylglycine
Assay Percent Range >98%
Molecular Formula C9H9NO5
Formula Weight 211.17
Accessories

Store at Room Temperature