Please login to your online account to display your discounted pricing

(R)-3-Carboxy-4-hydroxyphenylglycine, Tocris

Ionotropic glutamate receptor antagonist

$479.45 - $1,954.04

Chemical Identifiers

Chemical Name or Material (R)-3-Carboxy-4-hydroxyphenylglycine
Assay Percent Range >98%
Molecular Formula C9H9NO5
Formula Weight 211.17
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

032810

 
r & d systems
RDS032810
10mg Each for $479.45

032850

 
r & d systems
RDS032850
50mg Each for $1,954.04
Description & Specifications

Specifications

Quantity 10mg
Formula Weight 211.17
Chemical Name or Material (R)-3-Carboxy-4-hydroxyphenylglycine
Assay Percent Range >98%
CAS 13861-03-5
Molecular Formula C9H9NO5
Recommended Storage Store at Room Temperature
Safety and Handling

Store at Room Temperature