missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ (R)-3-(Boc-amino)butyric acid, 98%

$134.68 - $404.50

Chemical Identifiers

CAS 159991-23-8
Molecular Formula C9H17NO4
Molecular Weight (g/mol) 203.238
MDL Number MFCD00270343
InChI Key PYNDHEONPQYIAN-ZCFIWIBFSA-N
Synonym 3-n-boc-3-r-amino butyric acid, 3r-3-tert-butoxycarbonyl amino butanoic acid, 3r-3-tert-butoxycarbonylamino butanoic acid, boc-d-3-abu-oh, boc-d-3-aminobutyric acid, boc-d-beta-homoalanine, r-3-tert-butoxycarbonyl amino butanoic acid, r-3-tert-butoxycarbonylamino butanoic acid, r-3-tert-butoxycarbonylamino-butyric acid, r-n-boc-3-aminobutyric acid
PubChem CID 2761507
IUPAC Name (3R)-3-[(2-methylpropan-2-yl)oxycarbonylamino]butanoic acid
SMILES CC(CC(=O)O)NC(=O)OC(C)(C)C
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH62750MD View Documents Alfa Aesar™
H62750MD
250mg Each for $134.68
 
AAH6275003 View Documents Alfa Aesar™
H6275003
1g Each for $404.50
 
Description

Description

Applications
(R)-3-(Boc-amino)butyric acid is employed as a intermediate for pharmaceutical and chemical research.

Solubility
Slightly soluble in water.

Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
Specifications

Chemical Identifiers

159991-23-8
203.238
PYNDHEONPQYIAN-ZCFIWIBFSA-N
2761507
CC(CC(=O)O)NC(=O)OC(C)(C)C
C9H17NO4
MFCD00270343
3-n-boc-3-r-amino butyric acid, 3r-3-tert-butoxycarbonyl amino butanoic acid, 3r-3-tert-butoxycarbonylamino butanoic acid, boc-d-3-abu-oh, boc-d-3-aminobutyric acid, boc-d-beta-homoalanine, r-3-tert-butoxycarbonyl amino butanoic acid, r-3-tert-butoxycarbonylamino butanoic acid, r-3-tert-butoxycarbonylamino-butyric acid, r-n-boc-3-aminobutyric acid
(3R)-3-[(2-methylpropan-2-yl)oxycarbonylamino]butanoic acid

Specifications

250mg
C9H17NO4
3-n-boc-3-r-amino butyric acid, 3r-3-tert-butoxycarbonyl amino butanoic acid, 3r-3-tert-butoxycarbonylamino butanoic acid, boc-d-3-abu-oh, boc-d-3-aminobutyric acid, boc-d-beta-homoalanine, r-3-tert-butoxycarbonyl amino butanoic acid, r-3-tert-butoxycarbonylamino butanoic acid, r-3-tert-butoxycarbonylamino-butyric acid, r-n-boc-3-aminobutyric acid
CC(CC(=O)O)NC(=O)OC(C)(C)C
203.238
203.24
(R)-3-(Boc-amino)butyric acid
159991-23-8
MFCD00270343
PYNDHEONPQYIAN-ZCFIWIBFSA-N
(3R)-3-[(2-methylpropan-2-yl)oxycarbonylamino]butanoic acid
2761507
98%