missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ (R)-3-(Boc-amino)butyric acid, 98%

Manufacturer:  Alfa Aesar™ H62750MD

 View more versions of this product

Catalog No. AAH62750MD



Description

Description

Applications
(R)-3-(Boc-amino)butyric acid is employed as a intermediate for pharmaceutical and chemical research.

Solubility
Slightly soluble in water.

Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
Specifications

Specifications

(R)-3-(Boc-amino)butyric acid
159991-23-8
MFCD00270343
PYNDHEONPQYIAN-ZCFIWIBFSA-N
(3R)-3-[(2-methylpropan-2-yl)oxycarbonylamino]butanoic acid
2761507
98%
250mg
C9H17NO4
3-n-boc-3-r-amino butyric acid, 3r-3-tert-butoxycarbonyl amino butanoic acid, 3r-3-tert-butoxycarbonylamino butanoic acid, boc-d-3-abu-oh, boc-d-3-aminobutyric acid, boc-d-beta-homoalanine, r-3-tert-butoxycarbonyl amino butanoic acid, r-3-tert-butoxycarbonylamino butanoic acid, r-3-tert-butoxycarbonylamino-butyric acid, r-n-boc-3-aminobutyric acid
CC(CC(=O)O)NC(=O)OC(C)(C)C
203.238
203.24

Chemical Identifiers

159991-23-8
203.238
PYNDHEONPQYIAN-ZCFIWIBFSA-N
2761507
CC(CC(=O)O)NC(=O)OC(C)(C)C
C9H17NO4
MFCD00270343
3-n-boc-3-r-amino butyric acid, 3r-3-tert-butoxycarbonyl amino butanoic acid, 3r-3-tert-butoxycarbonylamino butanoic acid, boc-d-3-abu-oh, boc-d-3-aminobutyric acid, boc-d-beta-homoalanine, r-3-tert-butoxycarbonyl amino butanoic acid, r-3-tert-butoxycarbonylamino butanoic acid, r-3-tert-butoxycarbonylamino-butyric acid, r-n-boc-3-aminobutyric acid
(3R)-3-[(2-methylpropan-2-yl)oxycarbonylamino]butanoic acid
Documents