missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ (R)-3-Boc-4-methyl-2,2-dioxo-1,2,3-oxathiazolidine, 97%

Manufacturer:  Alfa Aesar™ H6388903

 View more versions of this product

Catalog No. AAH6388903



Description

Description

Applications
Thiazoles are used in peptide research. Mainly used in medicine field.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents. 2-7°C Refrigerate.
Specifications

Specifications

(R)-3-Boc-4-methyl-2,2-dioxo-1,2,3-oxathiazolidine
100°C to 107°C
1212136-00-9
MFCD09864994
RTHSATBSXDKZBN-QGZVFWFLSA-N
methyl (4R)-3-[bis(4-methoxyphenyl)methyl]-2,2-dioxooxathiazolidine-4-carboxylate
46172874
Solid
White to Brown
1g
C19H21NO7S
methyl r-3-bis 4-methoxyphenyl methyl-2,2-dioxo-1,2,3 oxathiazolidine-4-carboxylate, methyl 4r-3-bis 4-methoxyphenyl methyl-2,2-dioxo-1,2??,3-oxathiazolidine-4-carboxylate
COC1=CC=C(C=C1)C(C2=CC=C(C=C2)OC)N3C(COS3(=O)=O)C(=O)OC
407.437
237.27
97%

Chemical Identifiers

1212136-00-9
407.437
RTHSATBSXDKZBN-QGZVFWFLSA-N
46172874
COC1=CC=C(C=C1)C(C2=CC=C(C=C2)OC)N3C(COS3(=O)=O)C(=O)OC
C19H21NO7S
MFCD09864994
methyl r-3-bis 4-methoxyphenyl methyl-2,2-dioxo-1,2,3 oxathiazolidine-4-carboxylate, methyl 4r-3-bis 4-methoxyphenyl methyl-2,2-dioxo-1,2??,3-oxathiazolidine-4-carboxylate
methyl (4R)-3-[bis(4-methoxyphenyl)methyl]-2,2-dioxooxathiazolidine-4-carboxylate
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

storageNote1 : 2° to 8°C

SDS Documents