missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ (R)-(+)-3-Boc-2,2-dimethyloxazolidine-4-carboxaldehyde, 95%

Manufacturer:  Alfa Aesar™ H63351MD

 View more versions of this product

Catalog No. AAH63351MD



Description

Description

Applications
(R)-(+)-3-Boc-2,2-dimethyloxazolidine-4-carboxaldehyde is used in the preparation of fluorinated analogs of glutemic and glutamine.

Notes
Incompatible with strong oxidizing agents.
Specifications

Specifications

(R)-(+)-3-Boc-2,2-dimethyloxazolidine-4-carboxaldehyde
1.4450
67°C (0.3mmHg)
250mg
C11H19NO4
3651554
r-tert-butyl 4-formyl-2,2-dimethyloxazolidine-3-carboxylate, unii-upk623lv60, r-4-formyl-2,2-dimethyl-oxazolidine-3-carboxylic acid tert-butyl ester, tert-butyl 4r-4-formyl-2,2-dimethyl-1,3-oxazolidine-3-carboxylate, r-+-3-boc-2,2-dimethyloxazolidine-4-carboxaldehyde, tert-butyl r-+-4-formyl-2,2-dimethyl-3-oxazolidinecarboxylate, 3-oxazolidinecarboxylic acid, 4-formyl-2,2-dimethyl-, 1,1-dimethylethyl ester, 4r, r-garner aldehyde, r-3-boc-4-formyl-2,2-dimethyl-1,3-oxazolidine, r---3-boc-2,2-dimethyloxazolidine-4-carboxaldehyde
CC1(N(C(CO1)C=O)C(=O)OC(C)(C)C)C
229.276
229.27
95%
1.06g/mL
Air Sensitive
Colorless to Yellow
95715-87-0
MFCD00674064
14,4371
PNJXYVJNOCLJLJ-QMMMGPOBSA-N
tert-butyl (4R)-4-formyl-2,2-dimethyl-1,3-oxazolidine-3-carboxylate
178792
Liquid

Chemical Identifiers

95715-87-0
229.276
PNJXYVJNOCLJLJ-QMMMGPOBSA-N
178792
CC1(N(C(CO1)C=O)C(=O)OC(C)(C)C)C
C11H19NO4
MFCD00674064
r-tert-butyl 4-formyl-2,2-dimethyloxazolidine-3-carboxylate, unii-upk623lv60, r-4-formyl-2,2-dimethyl-oxazolidine-3-carboxylic acid tert-butyl ester, tert-butyl 4r-4-formyl-2,2-dimethyl-1,3-oxazolidine-3-carboxylate, r-+-3-boc-2,2-dimethyloxazolidine-4-carboxaldehyde, tert-butyl r-+-4-formyl-2,2-dimethyl-3-oxazolidinecarboxylate, 3-oxazolidinecarboxylic acid, 4-formyl-2,2-dimethyl-, 1,1-dimethylethyl ester, 4r, r-garner aldehyde, r-3-boc-4-formyl-2,2-dimethyl-1,3-oxazolidine, r---3-boc-2,2-dimethyloxazolidine-4-carboxaldehyde
tert-butyl (4R)-4-formyl-2,2-dimethyl-1,3-oxazolidine-3-carboxylate
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

storageNote1 : 2° to 8°C

SDS Documents