Please login to your online account to display your discounted pricing

Alfa Aesar™ (R)-(+)-3-(Benzyloxycarbonyl)oxazolidine-4-carboxylic acid, 98%

$85.74 - $85.74

Chemical Identifiers

Chemical Name or Material (R)-(+)-3-(Benzyloxycarbonyl)oxazolidine-4-carboxylic acid
CAS 97534-84-4
Molecular Formula C12H13NO5
Formula Weight 251.24
MDL Number MFCD00274192
Synonym N-Cbz-(R)-oxaproline; Z-D-OxaPro
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2724003

  SDS
alfa aesar™
H2724003
1g Each for $85.74
Description & Specifications

Specifications

Formula Weight 251.24
Chemical Name or Material (R)-(+)-3-(Benzyloxycarbonyl)oxazolidine-4-carboxylic acid
Assay 98%
Molecular Formula C12H13NO5
MDL Number MFCD00274192
CAS 97534-84-4
Synonym N-Cbz-(R)-oxaproline; Z-D-OxaPro
Melting Point 80° to 81°C
Quantity 1g
Optical Rotation +79° (c=1 in Chloroform)