missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

(R)-(+)-3-Benzyloxy-1,2-propanediol, TCI America™

$48.89 - $152.78

Chemical Identifiers

CAS 56552-80-8
Molecular Formula C10H14O3
Molecular Weight (g/mol) 182.219
MDL Number MFCD00067260
InChI Key LWCIBYRXSHRIAP-SNVBAGLBSA-N
Synonym (R)-(+)-Glycerol alpha-Benzyl Ether
PubChem CID 2724795
IUPAC Name (2R)-3-phenylmethoxypropane-1,2-diol
SMILES C1=CC=C(C=C1)COCC(CO)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
B21411G View Documents TCI America
B21411G
1g Each for $152.78
 
B2141100MG View Documents TCI America
B2141100MG
100mg Each for $48.89
 
Specifications

Chemical Identifiers

56552-80-8
182.219
LWCIBYRXSHRIAP-SNVBAGLBSA-N
2724795
C1=CC=C(C=C1)COCC(CO)O
C10H14O3
MFCD00067260
(R)-(+)-Glycerol alpha-Benzyl Ether
(2R)-3-phenylmethoxypropane-1,2-diol

Specifications

Yellow
56552-80-8
MFCD00067260
LWCIBYRXSHRIAP-SNVBAGLBSA-N
(2R)-3-phenylmethoxypropane-1,2-diol
2724795
Clear Liquid at 20°C
(R)-(+)-3-Benzyloxy-1,2-propanediol
1g
C10H14O3
(R)-(+)-Glycerol alpha-Benzyl Ether
C1=CC=C(C=C1)COCC(CO)O
182.219
182.22
≥98.0% (GC)
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Refrigerator