Learn More
(R)-(+)-3-Aminotetrahydrofuran p-toluenesulfonate salt, 95%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 435820050
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
(R)-(+)-3-Aminotetrahydrofuran p-toluenesulfonate salt | |
96.0 | |
48.91 to 52.99% C, 5.18 to 5.62% N | |
95% | |
C11H17NO4S | |
5g | |
r-3-aminotetrahydrofuran tosylate, r-+-tetrahydro-3-furylamine p-toluenesulfonate salt, r---3-aminotetrahydrofuran toluene-4-sulfonate, r-+-3-aminotetrahydrofuran toluene-4-sulfonate, r---3-aminotetrahydrofurantoluene-4-sulfonate, 3-furanamine, tetrahydro-, r-, 4-methylbenzenesulfonate, c4h9no.c7h8o3s, r-tetrahydrofuran-3-amine-4-methylbenzenesulfonate, 3r-oxolan-3-amine; para-toluene sulfonate, 4-methylbenzenesulfonic acid; 3r-tetrahydrofuran-3-amine | |
NC1CCOC1.CC1=CC=C(C=C1)S(O)(=O)=O | |
259.32 | |
259.32 |
111769-27-8 | |
100.0 | |
Authentic | |
Glass bottle | |
MFCD00040622,MFCD00040622,MFCD00066238 | |
4.3 | |
BZXPLADBSZWDIH-UHFFFAOYNA-N | |
4-methylbenzenesulfonic acid;(3R)-oxolan-3-amine | |
14243169 | |
95% |
Chemical Identifiers
111769-27-8 | |
259.32 | |
BZXPLADBSZWDIH-UHFFFAOYNA-N | |
14243169 | |
NC1CCOC1.CC1=CC=C(C=C1)S(O)(=O)=O |
C11H17NO4S | |
MFCD00040622,MFCD00040622,MFCD00066238 | |
r-3-aminotetrahydrofuran tosylate, r-+-tetrahydro-3-furylamine p-toluenesulfonate salt, r---3-aminotetrahydrofuran toluene-4-sulfonate, r-+-3-aminotetrahydrofuran toluene-4-sulfonate, r---3-aminotetrahydrofurantoluene-4-sulfonate, 3-furanamine, tetrahydro-, r-, 4-methylbenzenesulfonate, c4h9no.c7h8o3s, r-tetrahydrofuran-3-amine-4-methylbenzenesulfonate, 3r-oxolan-3-amine; para-toluene sulfonate, 4-methylbenzenesulfonic acid; 3r-tetrahydrofuran-3-amine | |
4-methylbenzenesulfonic acid;(3R)-oxolan-3-amine |
Safety and Handling
GHS H Statement
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF
GHS Signal Word: Warning
RUO â Research Use Only