Please login to your online account to display your discounted pricing

Alfa Aesar™ (R)-(-)-3-Aminopyrrolidine dihydrochloride, 98%

$71.58 - $71.58

Chemical Identifiers

Chemical Name or Material (R)-(-)-3-Aminopyrrolidine dihydrochloride
CAS 116183-81-4
Molecular Formula C4H10N2·2HCl
Formula Weight 159.06
MDL Number MFCD00070604
Beilstein 5852700
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL11173MD

 
alfa aesar™
L11173MD
250mg Each for $71.58
Description & Specifications

Specifications

Formula Weight 159.06
Chemical Name or Material (R)-(-)-3-Aminopyrrolidine dihydrochloride
Molecular Formula C4H10N2·2HCl
MDL Number MFCD00070604
CAS 116183-81-4
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Assay 98%
Beilstein 5852700
Melting Point ≈283°C
Quantity 250mg
Optical Rotation −1.2° (c=10 in Water)
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.