missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ (R)-(-)-3-Aminopyrrolidine dihydrochloride, 98%

Manufacturer:  Alfa Aesar™ L11173MD

 View more versions of this product

Catalog No. AAL11173MD



Specifications

Specifications

(R)-(-)-3-Aminopyrrolidine dihydrochloride
~283°C
116183-81-4
MFCD00070604
3r---3-aminopyrrolidine dihydrochloride, r---3-aminopyrrolidine dihydrochloride, r-pyrrolidin-3-amine dihydrochloride, r-3-aminopyrrolidine 2hcl, r-+-3-aminopyrrolidine 2hcl, 3r-pyrrolidin-3-amine dihydrochloride, 3r-3-pyrrolidinamine dihydrochloride, r-3-aminopyrrolidine dihydrochloride, 3-pyrrolidinamine, dihydrochloride, r, pubchem5730
C1CNCC1N.Cl.Cl
159.054
159.06
−1.2° (c=10 in Water)
250mg
C4H12Cl2N2
5852700
NJPNCMOUEXEGBL-RZFWHQLPSA-N
(3R)-pyrrolidin-3-amine;dihydrochloride
13500643
98%

Chemical Identifiers

116183-81-4
159.054
NJPNCMOUEXEGBL-RZFWHQLPSA-N
13500643
C1CNCC1N.Cl.Cl
C4H12Cl2N2
MFCD00070604
3r---3-aminopyrrolidine dihydrochloride, r---3-aminopyrrolidine dihydrochloride, r-pyrrolidin-3-amine dihydrochloride, r-3-aminopyrrolidine 2hcl, r-+-3-aminopyrrolidine 2hcl, 3r-pyrrolidin-3-amine dihydrochloride, 3r-3-pyrrolidinamine dihydrochloride, r-3-aminopyrrolidine dihydrochloride, 3-pyrrolidinamine, dihydrochloride, r, pubchem5730
(3R)-pyrrolidin-3-amine;dihydrochloride
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

Documents