Please login to your online account to display your discounted pricing

Alfa Aesar™ (R)-(-)-3-Aminopiperidine dihydrochloride, 98%

$64.80 - $166.24

Chemical Identifiers

Chemical Name or Material (R)-(-)-3-Aminopiperidine dihydrochloride
CAS 334618-23-4
Molecular Formula C5H12N2·2HCl
Formula Weight 173.08
MDL Number MFCD06799458
Synonym (R)-3-Piperidinamine dihydrochloride
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2768303

  SDS
alfa aesar™
H2768303
1g Each for $166.24

AAH27683MD

  SDS
alfa aesar™
H27683MD
250mg Each for $64.80
Description & Specifications

Specifications

Formula Weight 173.08
Chemical Name or Material (R)-(-)-3-Aminopiperidine dihydrochloride
Assay 98%
Molecular Formula C5H12N2·2HCl
MDL Number MFCD06799458
CAS 334618-23-4
Synonym (R)-3-Piperidinamine dihydrochloride
Melting Point ≈210°C
Health Hazard 1 Warning
Quantity 1g
Optical Rotation −1.8° (c=1 in Water)
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.