missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

(R)-3-Amino-3-phenylpropan-1-ol, 95%, 98% ee, ACROS Organics™

$1,225.46 - $1,225.46

Chemical Identifiers

CAS 170564-98-4
Molecular Formula C9H13NO
Molecular Weight (g/mol) 151.21
MDL Number MFCD01311791
InChI Key SEQXIQNPMQTBGN-SECBINFHSA-N
Synonym 3r-3-amino-3-phenylpropan-1-ol, pubchem13853, r--phenylalaninol, r-1-phenyl-3-propanolamine, r-3-amino-3-phenyl-1-propanol, r-3-amino-3-phenyl-propan-1-ol, r-3-amino-3-phenylpropan-1-ol, r-3-phenyl-beta-alaninol, r-beta-phenylalaninol
PubChem CID 7016858
IUPAC Name (3R)-3-amino-3-phenylpropan-1-ol
SMILES C1=CC=C(C=C1)C(CCO)N
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC363120010
SDS View Documents
Thermo Fisher Scientific
363120010
1g Glass bottle
Each for $1,225.46
 
Description

Description

Specifications

Chemical Identifiers

170564-98-4
151.21
SEQXIQNPMQTBGN-SECBINFHSA-N
7016858
C1=CC=C(C=C1)C(CCO)N
C9H13NO
MFCD01311791
3r-3-amino-3-phenylpropan-1-ol, pubchem13853, r--phenylalaninol, r-1-phenyl-3-propanolamine, r-3-amino-3-phenyl-1-propanol, r-3-amino-3-phenyl-propan-1-ol, r-3-amino-3-phenylpropan-1-ol, r-3-phenyl-beta-alaninol, r-beta-phenylalaninol
(3R)-3-amino-3-phenylpropan-1-ol

Specifications

97.5% min.
Glass bottle
1g
94.0
94% min. (GC)
MFCD01311791
SEQXIQNPMQTBGN-SECBINFHSA-N
(3R)-3-amino-3-phenylpropan-1-ol
7016858
Viscous Liquid or Low Melting Solid
(R)-3-Amino-3-phenylpropan-1-ol, 95%, (98% e.e.)
Authentic
White to Yellow or Yellow
170564-98-4
100.0
C9H13NO
3r-3-amino-3-phenylpropan-1-ol, pubchem13853, r--phenylalaninol, r-1-phenyl-3-propanolamine, r-3-amino-3-phenyl-1-propanol, r-3-amino-3-phenyl-propan-1-ol, r-3-amino-3-phenylpropan-1-ol, r-3-phenyl-beta-alaninol, r-beta-phenylalaninol
C1=CC=C(C=C1)C(CCO)N
151.21
151.21
95%,98% ee
Safety and Handling

Safety and Handling

GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

TSCA : TSCA

SDS
Product Certifications