missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

(R)-(-)-3-Amino-1-BOC-piperidine, 97%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC436490010

 View more versions of this product

Catalog No. AC436490010

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Specifications

Specifications

(R)-(-)-3-Amino-1-BOC-piperidine
Glass bottle
Colorless to Yellow
188111-79-7
100.0
C10H20N2O2
AKQXKEBCONUWCL-MRVPVSSYSA-N
tert-butyl (3R)-3-aminopiperidine-1-carboxylate
1501974
Liquid
Authentic
-30
1g
96.0
96% min. (GC)
1-piperidinecarboxylic acid, 3-amino-, 1,1-dimethylethyl ester, 3r, r-1-boc-3-amino-piperidine, r-1-boc-3-aminopiperidine, r-1-tert-butoxycarbonyl-3-aminopiperidine, r-3-amino-1-boc-piperidine, r-3-amino-1-n-boc-piperidine, r-3-amino-piperidine-1-carboxylic acid tert-butyl ester, r-tert-butyl 3-aminopiperidine-1-carboxylate, s-n-boc-3-aminopiperidine, tert-butyl 3r-3-aminopiperidine-1-carboxylate
CC(C)(C)OC(=O)N1CCCC(C1)N
200.28
200.28
97%

Chemical Identifiers

188111-79-7
200.28
1-piperidinecarboxylic acid, 3-amino-, 1,1-dimethylethyl ester, 3r, r-1-boc-3-amino-piperidine, r-1-boc-3-aminopiperidine, r-1-tert-butoxycarbonyl-3-aminopiperidine, r-3-amino-1-boc-piperidine, r-3-amino-1-n-boc-piperidine, r-3-amino-piperidine-1-carboxylic acid tert-butyl ester, r-tert-butyl 3-aminopiperidine-1-carboxylate, s-n-boc-3-aminopiperidine, tert-butyl 3r-3-aminopiperidine-1-carboxylate
tert-butyl (3R)-3-aminopiperidine-1-carboxylate
C10H20N2O2
AKQXKEBCONUWCL-MRVPVSSYSA-N
1501974
CC(C)(C)OC(=O)N1CCCC(C1)N
Safety and Handling

Safety and Handling

GHS H Statement
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF

GHS Signal Word: Warning

SDS Documents