Please login to your online account to display your discounted pricing

Alfa Aesar™ (R)-3-Acetylthiazolidine-4-carboxylic acid, 97%

Chemical Identifiers

Chemical Name or Material (R)-3-Acetylthiazolidine-4-carboxylic acid
CAS 54323-50-1
Formula Weight 175.2
MDL Number MFCD00038440
Beilstein 8389
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6544803

 
alfa aesar™
H6544803
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6544806

 
alfa aesar™
H6544806
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 175.2
Melting Point 143° to 145°C
Chemical Name or Material (R)-3-Acetylthiazolidine-4-carboxylic acid
Physical Form Solid
Assay 97%
MDL Number MFCD00038440
Beilstein 8389
CAS 54323-50-1
Storage Note 1 2° to 8°C
Boiling Point 439°C (760mmHg)
Density 1.46g/mL
Quantity 1g
Safety and Handling

2° to 8°C