Please login to your online account to display your discounted pricing

(R)-3,4-DCPG, Tocris

AMPA antagonist/weak NMDA antagonist

$252.39 - $1,050.42

Chemical Identifiers

Chemical Name or Material (R)-3, 4-DCPG
Assay Percent Range >96%
Molecular Formula C10H9NO6
Formula Weight 239.18
Synonym (R)-3, 4-Dicarboxyphenylglycine
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

139510

 
r & d systems
RDS139510
10mg Each for $252.39

139550

 
r & d systems
RDS139550
50mg Each for $1,050.42
Description & Specifications

Specifications

Formula Weight 239.18
Quantity 10mg
CAS 201730-10-1
Molecular Formula C10H9NO6
Assay Percent Range >96%
Synonym (R)-3, 4-Dicarboxyphenylglycine
Chemical Name or Material (R)-3, 4-DCPG
Recommended Storage Desiccate at Room Temperature
Safety and Handling

Desiccate at Room Temperature