Please login to your online account to display your discounted pricing

Alfa Aesar™ (R)-(-)-3,3-Dimethyl-2-butylamine, ChiPros, 99+%, ee 99+%

Chemical Identifiers

Chemical Name or Material (R)-(-)-3,3-Dimethyl-2-butylamine
CAS 66228-31-7
Molecular Formula C6H15N
Formula Weight 101.19
MDL Number MFCD00671643
UN Number UN2733
Beilstein 5237592
Synonym (R)-2-Amino-3,3-dimethylbutane
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1632603

  SDS
alfa aesar™
L1632603
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAL1632606

  SDS
alfa aesar™
L1632606
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAL1632614

  SDS
alfa aesar™
L1632614
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 101.19
CAS 66228-31-7
Health Hazard 1 Danger
Molecular Formula C6H15N
MDL Number MFCD00671643
Chemical Name or Material (R)-(-)-3,3-Dimethyl-2-butylamine
Synonym (R)-2-Amino-3,3-dimethylbutane
Assay Percent Notes ee ≥99%
Assay ≥99%
Health Hazard 3 GHS P Statement
P210-P260-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Flash Point 2°C (35°F)
DOT Information Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: AMINES, FLAMMABLE, CORROSIVE, N.O.S.
Health Hazard 2 GHS H Statement
H225-H314-H318
Highly flammable liquid and vapor.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Quantity 1g
Boiling Point 103°C
Beilstein 5237592
UN Number UN2733
Melting Point −4°C
Safety and Handling

Danger

GHS P Statement
P210-P260-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H225-H314-H318
Highly flammable liquid and vapor.
Causes severe skin burns and eye damage.
Causes serious eye damage.