Please login to your online account to display your discounted pricing

Alfa Aesar™ (R)-3,3'-Dibromo-2,2'-bis(methoxymethoxy)-1,1'-binaphthyl, 97%

$266.36 - $266.36

Chemical Identifiers

Chemical Name or Material (R)-3,3'-Dibromo-2,2'-bis(methoxymethoxy)-1,1'-binaphthyl
CAS 211734-49-5
Molecular Formula C24H20Br2O4
Formula Weight 532.23g/mol
MDL Number MFCD16294495
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH3155703

 
alfa aesar™
H31557-03
1g Each for $266.36
Description & Specifications

Specifications

Quantity 1g
Chemical Name or Material (R)-3,3'-Dibromo-2,2'-bis(methoxymethoxy)-1,1'-binaphthyl
CAS 211734-49-5
Assay 97%
Molecular Formula C24H20Br2O4
Formula Weight 532.23g/mol
MDL Number MFCD16294495